Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,6-Bis(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

10174-25G 10174-5G
76894-278EA 82.4 USD InStock
76894-278 76894-276
2,6-Bis(trifluoromethyl)benzoic acid ≥97%
2,6-Bis(trifluoromethyl)benzoic acid
Caution: for Research Use Only


Formula: C₉H₄F₆O₂
MW: 258.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00000376
CAS Number: 24821-22-5
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty