Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

H-D-Glu(OtBu)-OH 97%

Supplier: Ambeed
Warning
Synonyms: D-Glutamic acid 5-tert-butyl ester

A164413-25G A164413-1G A164413-10G
76973-728EA 91.67 USD InStock
76973-728 76973-730 76973-732
H-D-Glu(OtBu)-OH 97%
H-D-Glu(OtBu)-OH
Caution: for Research Use Only


Formula: C₉H₁₇NO₄
MDL Number: MFCD00153437
CAS Number: 45125-00-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty