Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cinnamaldehyde 98%

Supplier: Ambeed
Warning
Synonyms: 3-Phenylacrylaldehydetrans-Cinnamaldehyde

A331599-1000G A331599-25G A331599-500G A331599-100G
76978-260EA 5.15 USD InStock
76978-260 76978-262 76978-258 76978-256
Cinnamaldehyde 98%
Cinnamaldehyde
Caution: for Research Use Only


Formula: C₉H₈O
MW: 132.16 g/mol
MDL Number: MFCD00007000
CAS Number: 104-55-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty