Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Myristic acid 99%

Supplier: Ambeed
Warning
Synonyms: Tetradecylic acidTetradecanoic acid

A341595-100G A341595-5G A341595-1000G A341595-25G A341595-10G
76978-418EA 8.24 USD InStock
76978-418 76978-420 76978-416 76978-414
Myristic acid 99%
Myristic acid
Caution: for Research Use Only


Formula: C₁₄H₂₈O₂
MW: 228.38 g/mol
MDL Number: MFCD00002744
CAS Number: 544-63-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty