Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

p-Toluenesulfonyl isocyanate 99%

Supplier: Ambeed
Danger
Synonyms: 4-IsocyanatosulphonyltolueneTosyl isocyanate4-Isocyanatosulfonyltoluene

A404059-25G
76979-100EA 6.18 USD InStock
76979-100
p-Toluenesulfonyl isocyanate 99%
p-Toluenesulphonyl isocyanate
Caution: for Research Use Only


Formula: C₈H₇NO₃S
MW: 197.21 g/mol
MDL Number: MFCD00002030
CAS Number: 4083-64-1
UN: 2206
ADR: 6.1,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty