Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Phthaloylglycine 98%

Supplier: Ambeed
Warning
Synonyms: (1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)acetic acid

A111911-25G A111911-10G A111911-5G A111911-100G A111911-1G A111911-500G
76970-874EA 5.15 USD InStock
76970-874 76970-872 76970-878 76970-876 76970-880 76970-870
N-Phthaloylglycine 98%
N-Phthaloylglycine
Caution: for Research Use Only


Formula: C₁₀H₇NO₄
MW: 205.17 g/mol
MDL Number: MFCD00005900
CAS Number: 4702-13-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty