Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-aminopyrazine-2-carboxamide 95%

Supplier: Ambeed
Warning
Synonyms: 3-Aminopyrazine-2-carboxamide

A103087-1G
76970-256EA 86.52 USD InStock
76970-256
3-aminopyrazine-2-carboxamide 95%
3-aminopyrazine-2-carboxamide
Caution: for Research Use Only


Formula: C₅H₆N₄O
MW: 138.13 g/mol
MDL Number: MFCD03407446
CAS Number: 32587-10-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty