Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Palmitic acid 95%

Supplier: Ambeed
Warning
Synonyms: Hexadecanoic acidHexadecylic acid

A920864-1000G A920864-500G A920864-100G
76981-974EA 10.3 USD InStock
76981-974 76981-970 76981-972
Palmitic acid 95%
Palmitic acid
Caution: for Research Use Only


Formula: C₁₆H₃₂O₂
MW: 256.43 g/mol
MDL Number: MFCD00002747
CAS Number: 57-10-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty