Would you like to log in again?
Archimedes Principle Kit
Supplier: EISCO SCIENTIFIC LLC
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
PH0121
470344-540EA 29.75 USD InStock
470344-540
Archimedes Principle Kit
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
Calculate density the old school way, measuring the net force acting on a submerged object.
  • Compare the densities of two objects - one regularly shaped, and one irregular
  • Measure volume by the displacement method
  • Measure net force on the submerged object using a spring scale
Most of us have heard the apocryphal story of Archimedes running through the streets of Syracuse, but few actually understand Archimedes' Principle, that the buoyant force on an object submerged in a fluid is equal to the weight of the fluid that is displaced by that object.
This simple kit can be used to calculate the density of an irregularly shaped object. Using a spring scale and an overflow displacement vessel, students can calculate the bouyant force of two objects submersed in water. A couple of free body diagrams, and a sum of forces later, and physics students can calculate and compare the densities of the objects. Eureka!
Ordering information: Required but not included: Ring stand and clamp.
Delivery information: Kit includes: 2 metal solids, beaker, spring scale (N), and displacement vessel for water displacement experiments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty