Would you like to log in again?
Globe Glass™ Amber Media Bottles
Supplier: Globe Scientific
Globe Glass™
8110100 8110250 8110500 8112000 8110050 8111000
76683-672BX 425.09 USD InStock
76683-672 76683-670 76683-712 76683-732 76683-740 76683-696
Globe Glass™ Amber Media Bottles
Bottles Glass Bottles
Globe Glass™ amber media bottles are designed with a wide mouth for ease of filling, dispensing, and pipetting.
  • ASTM E438, Type I, Class A, borosilicate 3.3 glass
  • Amber color reduces light transmission
  • GlassGuard™ packaging system protects against transit damage
  • Pouring ring for drip-free liquid transfer
  • High resistance to chemicals
  • Dual scale metric graduations
  • Durable white enamel printing
A drip-free pour ring and autoclavable polypropylene (PP) screw cap enable sterilization of media and reagents. Globe Glass™ amber media bottles are a great choice for mixing, storing, and transporting light-sensitive solutions and media. Available in sizes ranging from 50 to 2000 ml.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty