Would you like to log in again?
PTS Panels® eGLU Test Strips
Supplier: PTS Diagnostics
PTS Panels®
2713
77020-352EA 69.64 USD InStock
77020-352
PTS Panels® eGLU Test Strips
Clinical Diagnostic Supplies Clinical Reagents and Kits
PTS Panels eGLU glucose test strips provide lab-quality glucose results on-site in seconds from a single fingerstick.
  • Measures glucose
  • 50 electrochemical test strips per vial
  • Room temperature storage
  • Available for CardioChek Plus analyzers only
  • Sample: 1.1 µl whole blood (fingerstick)
PTS Panels eGLU glucose test strips are for professional use exclusively with the CardioChek Plus analyzer to measure glucose in whole blood.
Glucose measurements are used in the diagnosis and treatment of carbohydrate metabolism disorders including diabetes mellitus, neonatal hypoglycemia, and idiopathic hypoglycemia, and of pancreatic islet carcinoma.
The test system will detect glucose levels from 40 to 600 mg/dl (2.22 to 33.3 mmol/L) and will display a number value for results in this range.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty