Would you like to log in again?
Globe Glass™ Griffin Style Beakers, Low Form, Heavy Duty
Supplier: Globe Scientific
Globe Glass™
8020600 8022000 8024000 8020400 8020250 8021000
76683-684BX 176.14 USD InStock
76683-684 76683-692 76683-718 76683-716 76683-710 76683-720
Globe Glass™ Griffin Style Beakers, Low Form, Heavy Duty
Beakers
Uniform wall thickness, flat bottoms and low expansion 3.3 Borosilicate glass make Globe Glass™ heavy duty, low form Griffin style beakers strong and durable.
  • Made exclusively from ASTM E438, Type I, Class A, 3.3 borosilicate glass
  • GlassGuard™ packaging system protects against transit damage
  • Dual metric scale graduations
  • Durable white enamel printing
  • Large white marking area
  • Formed pour spout for easy pouring
Thicker walls improve mechanical strength to protect against breakage even with the most rigorous use. Beaded rims and formed spouts facilitate accurate and steady pouring of liquids. Globe Glass low form Griffin style beakers are available in sizes from 250 to 4000 ml.
Certifications: Meet ASTM E960 Type II Specifications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty