Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

H-d-glu-otbu 97%

Supplier: Ambeed
Warning
Synonyms: D-Glutamic acid 1-tert-butyl esterH-D-Glu-OtBu

A104057-250MG A104057-1G
76994-720EA 13.39 USD InStock
76994-720 76994-718
H-d-glu-otbu 97%
H-d-glu-otbu
Caution: for Research Use Only


Formula: C₉H₁₇NO₄
MW: 203.238 g/mol
MDL Number: MFCD09842819
CAS Number: 25456-76-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty