Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride 95%

Supplier: Ambeed
Warning
Synonyms: L-Aspartatic Acid 4-Methyl Ester HydrochlorideH-Asp(OMe)-OH.HClH-Asp(OMe)-OH·HClL-Aspartic acid 4-methyl ester hydrochloride

A113763-10G
76995-372EA 7.21 USD InStock
76995-372
(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride 95%
(S)-2-Amino-4-methoxy-4-oxobutanoic acid hydrochloride
Caution: for Research Use Only


Formula: C₅H₉NO₄·HCl
MW: 183.6 g/mol
MDL Number: MFCD00038972
CAS Number: 16856-13-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty