Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

tert-Butyl-3-hydroxyphenylcarbamate 98%

Supplier: Ambeed
Warning
Synonyms: tert-Butyl (3-hydroxyphenyl)carbamate

A168384-5G A168384-1G A168384-25G
76998-114EA 92.7 USD InStock
76998-114 76998-116 76998-112
tert-Butyl-3-hydroxyphenylcarbamate 98%
tert-Butyl-3-hydroxyphenylcarbamate
Caution: for Research Use Only


Formula: C₁₁H₁₅NO₃
MW: 209.25 g/mol
MDL Number: MFCD06798070
CAS Number: 19962-06-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty