Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl-3-(trifluoromethyl)benzoate 98%

Supplier: Ambeed
Danger
Synonyms: Methyl 3-(trifluoromethyl)benzoate

A228600-10G A228600-25G A228600-5G
77000-122EA 10.3 USD InStock
77000-122 77000-124 77000-126
Methyl-3-(trifluoromethyl)benzoate 98%
Methyl 3-(trifluoromethyl)benzoate
Caution: for Research Use Only


Formula: C₉H₇F₃O₂
MW: 204.15 g/mol
MDL Number: MFCD00043543
CAS Number: 2557-13-3
UN: 3272
ADR: 3,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty