Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-γ-Undecalactone 98%

Supplier: Ambeed
Warning
Synonyms: Undecanoic gamma-Lactone(±)-Undecan-4-olide

A269485-500G A269485-25G A269485-100G
77001-128EA 6.18 USD InStock
77001-128 77001-126 77001-130
(±)-γ-Undecalactone 98%
(±)-γ-Undecalactone
Caution: for Research Use Only


Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
MDL Number: MFCD00005405
CAS Number: 104-67-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty