Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Chloro-1H-indole-3-carbaldehyde 97%

Supplier: Ambeed
Warning
Synonyms: 6-Chloroindole-3-carboxaldehyde

A283527-25G A283527-100G A283527-1G
77032-192EA 8.24 USD InStock
77032-192 77032-194 77032-190
6-Chloro-1H-indole-3-carbaldehyde 97%
6-Chloro-1H-indole-3-carbaldehyde
Caution: for Research Use Only


Formula: C₉H₆ClNO
MW: 179.60 g/mol
MDL Number: MFCD06657152
CAS Number: 703-82-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty