Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4,6-Triisopropylbenzoic acid 97%

Supplier: Ambeed
Warning

A320999-5G A320999-25G A320999-10G
77032-776EA 71.07 USD InStock
77032-776 77032-772 77032-774
2,4,6-Triisopropylbenzoic acid 97%
2,4,6-Triisopropylbenzoic acid
Caution: for Research Use Only


Formula: C₁₆H₂₄O₂
MW: 248.37 g/mol
MDL Number: MFCD00015031
CAS Number: 49623-71-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty