Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boc-D-Glu(Obzl)-OH 96%

Supplier: Ambeed
Warning
Synonyms: Boc-D-Glu(OBzl)-OHN-Boc-D-glutamic acid 5-benzyl ester

A333428-5G A333428-1G A333428-250MG
77032-942EA 20.6 USD InStock
77032-942 77032-940 77032-938
Boc-D-Glu(Obzl)-OH 96%
Boc-D-Glu(Obzl)-OH
Caution: for Research Use Only


Formula: C₁₇H₂₃NO₆
MW: 337.372 g/mol
MDL Number: MFCD00038258
CAS Number: 35793-73-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty