Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(1H-Indol-2-yl)methanamine 98%

Supplier: Ambeed
Warning

A377323-1G A377323-250MG A377323-100MG
77016-028EA 9.27 USD InStock
77016-028 77016-032 77016-030
(1H-Indol-2-yl)methanamine 98%
(1H-Indol-2-yl)methanamine
Caution: for Research Use Only


Formula: C₉H₁₀N₂
MW: 146.19 g/mol
MDL Number: MFCD03422512
CAS Number: 21109-25-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty