Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(tert-Butoxycarbonyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A401485-250MG A401485-5G
77003-916EA 97.85 USD InStock
77003-916 77003-914
1-(tert-Butoxycarbonyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylic acid 95%
1-(tert-Butoxycarbonyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole-3-carboxylic acid
Caution: for Research Use Only


Formula: C₁₇H₂₀N₂O₄
MW: 316.35 g/mol
MDL Number: MFCD29916819
CAS Number: 1910779-16-6
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty