Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-N-fmoc-L-phenylalanine 98%

Supplier: Ambeed
Warning
Synonyms: Fmoc-Phe(2-Br)-OH

A842116-5G A842116-1G
77036-644EA 21.63 USD InStock
77036-644 77036-646
2-Bromo-N-fmoc-L-phenylalanine 98%
2-Bromo-N-fmoc-L-phenylalanine
Caution: for Research Use Only


Formula: C₂₄H₂₀BrNO₄
MW: 466.331 g/mol
MDL Number: MFCD01311743
CAS Number: 220497-47-2
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty