Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Adipic acid 95%

Supplier: Ambeed
Warning
Synonyms: 1,6-Hexanedioic acidHexanedioc acid

A855080-100G A855080-25G
77006-848EA 6.18 USD InStock
77006-848 77006-850
Adipic acid 95%
Adipic acid
Caution: for Research Use Only


Formula: HOOC(CH₂)₄COOH
MW: 146.14 g/mol
MDL Number: MFCD00004420
CAS Number: 124-04-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty