Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-Penicillamine 95%

Supplier: Ambeed
Danger
Synonyms: 2-Amino-3-mercapto-3-methylbutanoic acid

A863168-250MG A863168-100MG A863168-5G
76993-668EA 5.15 USD InStock
76993-668 76993-672 76993-670
DL-Penicillamine 95%
DL-Penicillamine
Caution: for Research Use Only


Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
MDL Number: MFCD00004856
CAS Number: 52-66-4
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty