Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Amino-1H-1,2,4-triazole-5-thiol 95%

Supplier: Ambeed
Warning
Synonyms: 3-Amino-5-mercapto-1,2,4-triazole

A919564-10G A919564-25G A919564-500G
77007-102EA 8.24 USD InStock
77007-102 77007-104 77007-100
3-Amino-1H-1,2,4-triazole-5-thiol 95%
3-Amino-1H-1,2,4-triazole-5-thiol
Caution: for Research Use Only


Formula: C₂H₄N₄S
MW: 116.15 g/mol
MDL Number: MFCD00005231
CAS Number: 16691-43-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty