Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Denosumab Mouse Monoclonal Antibody [clone: 9A10]

Supplier: Genscript
GenScript®
A02102-40
77007-478EA 150.88 USD InStock
77007-478
Anti-Denosumab Mouse Monoclonal Antibody [clone: 9A10]
Antibodies
Denosumab with trade name Prolia, is an FDA-approved drug for the treatment of patients with osteoporosis, treatment-induced bone loss, metastases to bone, and giant cell tumor of bone. Denosumab is a fully human monoclonal antibody that works as a RANK ligand (RANKL) inhibitor. Denosumab can bind RANKL, preventing RANKL from activating RANK, its receptor on the osteoclast surface. Anti-Denosumab Antibody (9A10), mAb, Mouse is produced from a hybridoma resulting from the fusion of partner and B-lymphocytes obtained from a mouse immunized with Denosumab.

  • High specificity
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.

Type:
Antigen: Denosumab
Clonality: Monoclonal
Clone: 9A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty