Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Human CD8 Mouse Monoclonal Antibody

Supplier: Genscript
GenScript®
A02212-100
77007-482EA 52.25 USD InStock
77007-482
Anti-Human CD8 Mouse Monoclonal Antibody
Antibodies
CD8 (cluster of differentiation 8) is a member of the immunoglobulin superfamily found on the majority of thymocytes, a subset of peripheral blood T cells, and NK cells (which express almost exclusively CD8a homodimers). CD8 antigen is existed as a homodimer composed of two alpha chains or as a heterodimer composed of one alpha and one beta chain. CD8 acts as a co-receptor with MHC class I-restricted T cell receptors in antigen recognition and T cell activation and has been shown to play a role in thymic differentiation. CD8+ cytotoxic T cells are crucial for the killing of virus-infected cells and tumor cells.

  • GMP-grade anti-human
  • CD antibodies available
  • Fully validated performance in ex vivo cell-based assay
  • High bioactivity and lot-to-lot consistency
  • Animal-free manufacturing process

Anti-CD antibodies which specifically recognize CD proteins have many applications, including cell activation, cell type detection or identification, and cell sorting and isolation. GenScript offers an expanding selection of high-quality anti-CD antibodies to popular CD markers.

Type:
Antigen: Human CD8
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty