Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Eculizumab Mouse Monoclonal Antibody [clone: 4F6]

Supplier: Genscript
GenScript®
A02115-40
77007-628EA 150.88 USD InStock
77007-628
Anti-Eculizumab Mouse Monoclonal Antibody [clone: 4F6]
Antibodies
Eculizumab with trade name Soliris, is an FDA-approved drug to treat paroxysmal nocturnal hemoglobinuria (PNH), atypical hemolytic uremic syndrome (aHUS), and neuromyelitis optica. Eculizumab is a recombinant humanized monoclonal antibody against the complement protein C5. Eculizumab specifically binds to the terminal complement component 5, or C5. Binding to this protein prevents the activation of a complement terminal complex, which is used to treat a number of autoimmune conditions. Anti-Eculizumab Antibody (4F6), mAb, Mouse is produced from a hybridoma resulting from the fusion of partner and B-lymphocytes obtained from a mouse immunized with Eculizumab.

  • High specificity
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.

Type:
Antigen: Eculizumab
Clonality: Monoclonal
Clone: 4F6
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty