Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Ustekinumab Mouse Monoclonal Antibody [clone: 11E3]

Supplier: Genscript
GenScript®
A02119-40
77007-642EA 150.88 USD InStock
77007-642
Anti-Ustekinumab Mouse Monoclonal Antibody [clone: 11E3]
Antibodies
Ustekinumab with trade name Stelara, is an FDA-approved drug to Crohn's disease, Ulcerative Colitis, Plaque Psoriasis and Psoriatic Arthritis, targeting both interleukins (IL)-12 and IL-23. Ustekinumab is a fully human immunoglobulin G1 kappa mAb that binds to IL-12 and IL-23. Ustekinumab binds to the p40 subunit common to IL-12 and IL-23 and prevents their interaction with the IL-12 receptor β1 subunit of the IL-12 and IL-23 receptor complexes, thereby disrupting the proinflammatory pathway. Anti-Ustekinumab Antibody (11E3), mAb, Mouse is produced from a hybridoma resulting from the fusion of partner and B-lymphocytes obtained from a mouse immunized with Ustekinumab.

  • High specificity
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.

Type:
Antigen: Ustekinumab
Clonality: Monoclonal
Clone: 11E3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty