Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Pertuzumab Mouse Monoclonal Antibody [clone: 1A3]

Supplier: Genscript
GenScript®
A02150-40
77007-574EA 150.88 USD InStock
77007-574
Anti-Pertuzumab Mouse Monoclonal Antibody [clone: 1A3]
Antibodies
Pertuzumab (also called 2C4, trade name Perjeta) is a monoclonal antibody used in combination with trastuzumab and docetaxel for the treatment of metastatic HER2-positive breast cancer; it is also used in the same combination as a neoadjuvant in early HER2-positive breast cancer. The epitope for pertuzumab is the domain of HER2 where it binds to HER3, and pertuzumab prevents the HER2/HER3 dimer from forming, which in turn blocks signaling by the dimer and as a result prevents HER2 from functioning. Anti-Pertuzumab Antibody (1A3), mAb, Mouse is produced from a hybridoma resulting from the fusion of partner and B-lymphocytes obtained from a mouse immunized with Pertuzumab.

  • High specificity
  • High affinity
  • High stability

GenScript has developed a broad range of off-the-shelf anti-idiotype antibodies against blockbuster drugs to help accelerate your research of biosimilars.

Type:
Antigen: Pertuzumab
Clonality: Monoclonal
Clone: 1A3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty