Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-VHH Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 45H8]

Supplier: Genscript
GenScript®
A02172-200
77007-480EA 224.3 USD InStock
77007-480
Anti-VHH Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 45H8]
Antibodies
A camelid single-domain antibody (sdAb, called Nanobody by Ablynx, the developer) is a peptide chain of about 110 amino acids long, comprising one variable domain of a heavy-chain antibody, these are called VHH fragments. Like a whole antibody, it is able to bind selectively to a specific antigen. Single-domain antibodies allow a broad range of applications in biotechnical as well as therapeutic uses due to their small size, simple production, and high affinity. However, their sequences may increase the risk of immunogenicity and anti-drug antibody (ADA) development in humans, and thus, sdAbs are routinely humanized during development.

  • MonoRab™ technology guarantees high affinity: Kd ~10 to 11 M
  • No cross reactivity with other species
  • Specific to camelid IgG2 and IgG3
  • Recognize conformational epitopes of the VHH domain
  • Recognize both camelid and humanized VHHs

GenScript has developed a series of rabbit monoclonal (MonoRab) anti-VHH antibodies to facilitate your research of VHHs. Our anti-VHH antibodies recognize conformational epitopes of the VHH domain of camelid lgG2 and lgG3 subclasses. It is not necessary to add a tag to VHH for detection anymore.

Type:
Antigen: VHH
Clonality: Monoclonal
Clone: 45H8
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty