Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Human CD4 Mouse Monoclonal Antibody

Supplier: Genscript
GenScript®
A02211-100
77007-548EA 52.25 USD InStock
77007-548
Anti-Human CD4 Mouse Monoclonal Antibody
Antibodies
CD4 (cluster of differentiation 4) is a single glycoprotein with a molecular weight of 59 kDa. CD4 antigen is expressed on the surface of T helper cells, monocytes and macrophages. CD4 recognizes antigens associated with MHC class II molecules, and participates in cell-cell interactions, thymic differentiation, and signal transduction. CD4 acts as a primary receptor for HIV, binding to HIV gp120. CD4+ T helper cells bind and activate B cells to create immunoglobulins.

  • GMP-grade anti-human
  • CD antibodies available
  • Fully validated performance in ex vivo cell-based assay
  • High bioactivity and lot-to-lot consistency
  • Animal-free manufacturing process

Anti-CD antibodies which specifically recognize CD proteins have many applications, including cell activation, cell type detection or identification, and cell sorting and isolation. GenScript offers an expanding selection of high-quality anti-CD antibodies to popular CD markers.

Type:
Antigen: Human CD4
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty