Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Human CD28 Mouse Monoclonal Antibody

Supplier: Genscript
GenScript®
A02213-100
77007-618EA 52.25 USD InStock
77007-618
Anti-Human CD28 Mouse Monoclonal Antibody
Antibodies
CD28 (Cluster of Differentiation 28) is one of the proteins expressed on T cells that provide co-stimulatory signals required for T cell activation and survival. It plays an important role in cell proliferation and cytokine production and promotion of T-cell survival. It also enhances the production of IL-4 and IL-10 in T cells in conjunction with TCR/CD3 ligation and CD40L co-stimulation.

  • GMP-grade anti-human
  • CD antibodies available
  • Fully validated performance in ex vivo cell-based assay
  • High bioactivity and lot-to-lot consistency
  • Animal-free manufacturing process

Anti-CD antibodies which specifically recognize CD proteins have many applications, including cell activation, cell type detection or identification, and cell sorting and isolation. GenScript offers an expanding selection of high-quality anti-CD antibodies to popular CD markers.

Type:
Antigen: Human CD28
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty