Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Chloro-2-fluoroaniline 98%

Supplier: Ambeed
Warning

A189438-10G A189438-500G A189438-25G A189438-1000G A189438-100G
77084-552EA 6.18 USD InStock
77084-552 77274-390 77274-392 77274-386 77274-388
3-Chloro-2-fluoroaniline 98%
3-Chloro-2-fluoroaniline
Caution: for Research Use Only


Formula: C₆H₅ClFN
MW: 145.56 g/mol
MDL Number: MFCD00069415
CAS Number: 2106-04-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty