Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chlorobenzothiazole 98%

Supplier: Ambeed
Warning

A199145-5G A199145-10G A199145-100G A199145-500G A199145-25G
77139-292EA 187.46 USD InStock
77139-292 77139-294 77085-738 77139-290 77139-288
2-Chlorobenzothiazole 98%
2-Chlorobenzothiazole
Caution: for Research Use Only


Formula: C₇H₄ClNS
MW: 169.63 g/mol
MDL Number: MFCD00005776
CAS Number: 615-20-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty