Would you like to log in again?
Accucold® Vaccine Freezers, ADA Height
Supplier: Felix Storch Inc
Accucold®
AFZ2PVBIADA AFZ5PVBIADA
77041-314EA 4718.43 USD InStock
77041-314 77041-312
Accucold® Vaccine Freezers, ADA Height
Freezers Laboratory / General Purpose Freezers
Purpose-built all-freezers for vaccine storage in medical, pharmaceutical and life science applications.
  • Intelligent microprocessor digital temperature controller
  • Improved temperature accuracy
  • Powder-coated handle with naturally occurring silver ions to help reduce germ spread
  • Additional sensor reads interior air temperature for superior temperature control
Accucold's Pharma-Vac Performance series are upright freezers that are designed and purpose-built for pharmacy, medication, and vaccine applications to support meeting CDC/VFC guidelines.
All-freezer with a temperature range of −30 to −10 °C as recommended for a variety of vaccines and pharmaceuticals. Designed for freestanding or recessed installation and sized just under 32" high to fit under lower counters complying with ADA guidelines for accessibility. The microprocessor temperature controller is externally located to minimize door openings, helping to protect stored contents from ambient temperature. The buffered temperature probe is encased in a glycol-filled bottle to better simulate temperature of the stored product, with an additional sensor that reads air temperature to ensure superior temperature control. The current and min/max temperature display is viewable in Celsius or Fahrenheit to the nearest tenth of a degree. The audible and visual temperature alarm sounds if the unit's interior goes out of range. Additional alarms include a power failure alert, door open alarm, and sensor failure alarm. This model is equipped with remote alarm contacts in the rear and a ¹/₂" probe port on the right side to accept additional monitoring devices. A hospital grade cord with 'green dot' plug is also included for added safety in high traffic areas.
Inside, this freezer features adjustable plastic-coated wire shelves that can be positioned at ¹/₂" increments to accommodate virtually any sized item. This unit includes LED internal lighting with an on/off rocker switch. The interior is constructed from white powder coated metal to ensure added durability and better temperature retention. Static manual defrost cooling helps to ensure stable temperature conditions.
White cabinet finish with a blue accented control panel. The factory-installed antimicrobial handle is powder-coated with naturally occurring silver ions, which help to reduce the spread of germs, allowing for a safer, cleaner user experience. The user-reversible door ships with a right hand swing and self-closing function, while a factory-installed lock provides security as needed, with two keys for staff use. Powerful magnetic gaskets ensure a positive seal.
Designed with hydrocarbon refrigerant, Accucold Performance series freezers provide an energy efficient solution to vaccine storage. This unit is Pyxis, Omnicell, and AcuDose RX compatible. Additional sizes, as well as solid door options, are also available.
NOTE: Accucold freezers are purpose-built to the support meeting the current CDC (Centers for Disease Control) guidelines for the storage of vaccines. For additional temperature assurance, the CDC recommends a calibrated digital data logger (DDL), which can be added with the factory-installed option DL2B or addition of the DL2BKit accessory.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty