Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Dichlorobenzyl alcohol 98%

Supplier: Ambeed
Warning
Synonyms: (2,4-Dichlorophenyl)methanol

A116769-5G A116769-10G A116769-25G
77094-256EA 8.24 USD InStock
77094-256 77094-258 77094-254
2,4-Dichlorobenzyl alcohol 98%
2,4-Dichlorobenzyl alcohol
Caution: for Research Use Only


Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
MDL Number: MFCD00004606
CAS Number: 1777-82-8
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty