Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-2-fluorobenzoyl chloride 98%

Supplier: Ambeed
Danger
Synonyms: 4-Bromo-2-fluorobenzene-1-carbonyl chloride

A170860-5G A170860-1G A170860-25G
77096-502EA 18.54 USD InStock
77096-502 77096-506 77096-504
4-Bromo-2-fluorobenzoyl chloride 98%
4-Bromo-2-fluorobenzoyl chloride
Caution: for Research Use Only


Formula: C₇H₃BrClFO
MDL Number: MFCD03411584
CAS Number: 151982-51-3
UN: 3265
ADR: 8,Ⅱ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty