Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromobenzyl alcohol 98%

Supplier: Ambeed
Warning
Synonyms: (3-Bromophenyl)methanol

A218176-500G A218176-100G A218176-25G
77097-890EA 37.08 USD InStock
77097-890 77097-892 77303-482
3-Bromobenzyl alcohol 98%
3-Bromobenzyl alcohol
Caution: for Research Use Only


Formula: C₇H₇BrO
MW: 187.04 g/mol
MDL Number: MFCD00004629
CAS Number: 15852-73-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty