Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

DL-Valine 98%

Supplier: Ambeed
Warning
Synonyms: (±)-ValineH-DL-Val-OH2-Amino-3-methylbutanoic acid

A321883-25G A321883-500G A321883-100G
77099-856EA 6.18 USD InStock
77099-856 77099-858 77099-854
DL-Valine 98%
DL-Valine
Caution: for Research Use Only


Formula: C₅H₁₁NO₂
MW: 117.15 g/mol
MDL Number: MFCD00004267
CAS Number: 516-06-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty