Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluoro-N-methylbenzylamine 98%

Supplier: Ambeed
Warning
Synonyms: N-Methyl-4-fluorobenzylamine

A380568-1G A380568-5G
77101-690EA 47.38 USD InStock
77101-690 77101-688
4-Fluoro-N-methylbenzylamine 98%
4-Fluoro-N-methylbenzylamine
Caution: for Research Use Only


Formula: C₈H₁₀FN
MW: 139.17 g/mol
MDL Number: MFCD03839848
CAS Number: 405-66-3
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty