Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromo-1,2-difluorobenzene 98%

Supplier: Ambeed
Danger
Synonyms: 1-Bromo-3,4-difluorobenzene

A836724-5G A836724-25G
77105-856EA 5.15 USD InStock
77105-856 77105-854
4-Bromo-1,2-difluorobenzene 98%
4-Bromo-1,2-difluorobenzene
Caution: for Research Use Only


Formula: C₆H₃BrF₂
MW: 192.99 g/mol
MDL Number: MFCD00000304
CAS Number: 348-61-8
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty