Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Estes Rocket Science Starter Set

Supplier: ESTES INDUSTRIES INC SE
Estes
5302
470343-136EA 71.6 USD InStock
470343-136
Estes Rocket Science Starter Set
Educational Materials Physics Educational Materials Mechanics Learning Activities
The most complete model rocketry kit on the market.

  • Beginner-level build
  • Recommended engines: A8-3, B4-4, B6-4, C6-5
  • Max. altitude 335 m (1100')
  • Launch system, clinometer, and engines included

Experience the thrill of rocketry with the Crossfire™ ISX - a kit that is perfect for beginners and sure to impress everyone with its high-flying performance and sleek design! Are you ready for a high-flying adventure with the 'Rocket Science' model rocket kit? It has everything you need to launch and track your rocket! This kit is the perfect way to have fun while exploring STEM concepts. Using Estes standard model rocket engines (included with the kit), your rocket can reach impressive heights of up to 1100 ft.! Moreover, with the mini AltiTrak altitude tracker also included, you can measure and track its progress over multiple launches.

However, this kit isn't just about the excitement of launching rockets. It's also a fun and educational tool for learning about physics and engineering principles as you experiment with different engines and observe the reactions during launch. Whether you're a budding 'Rocket Scientist' or just looking for a unique way to have fun with STEM, the 'Rocket Science' model rocket kit is a fantastic choice.

Ordering information: Required but not included: (4) AA batteries.

Delivery information: Kit includes (1) crossfire ISX model rocket with parachute recovery, (1) Porta pad II, (1) electron beam launch controller, (1) mini AltiTrack, (1) B6-4 rocket engine, (1) C6-5 rocket engine, starters, and recovery wadding.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty