Would you like to log in again?
Wards® Forming Red, White, and Blue Chemical Demonstration Kit
Supplier: ALDON CORP SE
Wards®
IS7036
470337-952EA 34.65 USD InStock
470337-952
Wards® Forming Red, White, and Blue Chemical Demonstration Kit
Educational Materials Chemistry Educational Materials Ionic Properties Learning Activities
Discover the different chemical reaction types via this colorful demonstration.
  • Contains enough materials for 7 demonstrations
Introduce chemical reaction types. This is a patriotic demonstration that illustrates complex ion formation, double replacement while introducing chemical reaction types.
Delivery information: Includes 1×30 ml Iron (III) chloride, 1.0 M solution; 1×140 ml Silver nitrate, 0.1 M solution; 1×140 ml Potassium ferrocyanide, 0.1 M solution; 1×140 ml Potassium thiocyanate, 0.002 M solution.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty