Would you like to log in again?
Wards® Formation of Eutectic Alloys Chemical Demonstration
Supplier: ALDON CORP SE
Wards®
IS7014
470337-932EA 33.95 USD InStock
470337-932
Wards® Formation of Eutectic Alloys Chemical Demonstration
Educational Materials Chemistry Educational Materials Properties of Matter Learning Activities in Chemistry
Form a metal alloy by heating to metals to their eutectic point!
  • Produce a bismuth-tin alloy
Form a metal alloy as bismuth and tin are heated to the eutectic point of the two metals which is lower than the melting point of either metal.
Delivery information: Includes 1×100 ml Glycerin (Glycerol); 1×50 g Bismuth metal lumps; 1×15 g Tin metal mossy.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty