Would you like to log in again?
VirusGEN® LV Transfection Kit
Supplier: Mirus Bio
VirusGEN®
MIR6760
76501-634EA 1420.93 USD InStock
76501-634
VirusGEN® LV Transfection Kit
Transfection Reagents
TransIT-VirusGEN® transfection reagent is designed to enhance delivery of packaging and transfer plasmids to adherent and suspension HEK 293 cell types to increase recombinant adeno-associated virus (AAV) and lentivirus production.
  • Performance - efficient DNA delivery for production of high-titer viral vectors
  • Scalability - efficient production from <1 ml to large scale cultures
  • Flexibility - compatible with different recombinant virus and cell culture systems
  • Animal origin free - fully synthetic transfection reagent formulation
TransIT-VirusGEN® is ideal for academic research and biopharmaceutical discovery. Titers can be further increased with optimized enhancers included in the VirusGEN AAV Transfection Kit and VirusGEN LV transfection kit.
Ordering information: Kit includes: TransIT-VirusGEN transfection reagent (2×1.5 ml), VirusGEN LV complex formation solution and VirusGEN LV enhancer (100 ml).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty