Would you like to log in again?
Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit
Supplier: AAT Bioquest
22323
76779-088EA 537.56 USD InStock
76779-088
Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit
Assays Cellular Assays
It is sensitive and might be used for measuring active DNA synthesis at single-cell level.
  • blue laser compatible
  • highly reliable
  • optimized for flow cytometry
Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit uses XdU which is incorporated into cellular DNA during DNA synthesis.
After fixing cells, the incorporated XdU is labelled with iFluor® 488 MTA. The resulted iFluor® 488-labeled DNA formed in cells is visualized in FITC Channel. Bucculite™ Flow Cytometric XdU Cell Proliferation Assay Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry assay.
Kit provides an alternative to anti-BrdU antibody-based assay and EdU click chemistry assay
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty