Would you like to log in again?
PIP® Anser™ Semi-Rimless Safety Glasses
Supplier: Protective Industrial Products
PIP®
250-AN-10520
76944-628CS 751.3 USD InStock
76944-628
PIP® Anser™ Semi-Rimless Safety Glasses
Safety Glasses
Semi-rimless safety glasses feature a black frame, clear lens, and FogLess® 3Sixty™ coating.
  • Lightweight, semi-rimless design provides all day user comfort
  • Bayonet temples with flexible rubber tips offer a comfortable, secure fit
  • Soft rubber nose pads help keep the glasses in place
  • Supplied with an adjustable neck cord
  • 9 Base curve lens
Sleek, sporty style with wrap-around lens protection. The FogLess® 3Sixty™ enhanced hydrophobic coating resists lens fogging and provides clear vision in extreme working conditions.
Certifications: Meets ANSI Z87.1+.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty