Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chlorostyrene 97% stabilized

Supplier: Ambeed
Danger
Synonyms: 1-Chloro-2-vinylbenzene

Stabilization: Stabilised with hydroquinone (HQ)

A105945-10G A105945-5G A105945-1G
77107-634EA 26.78 USD InStock
77107-634 77107-638 77107-636
2-Chlorostyrene 97% stabilized
2-Chlorostyrene
Caution: for Research Use Only


Formula: C₈H₇Cl
MW: 138.6 g/mol
MDL Number: MFCD00000567
CAS Number: 2039-87-4
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty